Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-KLVFFA | 342877-55-8 | 99.26% | 723.90 | 5 MG
SDP

Catalog No. 5000398238
Encompass_Preferred

D-KLVFFA is an inhibitor of Amyloid-β assembly, with an IC50 of 2.6 μM, that can be utilized in Alzheimer's disease studies. This peptide is provided as a white to off-white solid with high purity, making it suitable for various research applications.

  • Inhibitor of Amyloid-β assembly
  • IC50 of 2.6 μM
  • Useful for Alzheimer's disease studies
  • Provided as a white to off-white solid
  • Purity of 99.26%
  • Molecular formula C38H57N7O7
  • Molecular weight of 723.90

Catalog No. 50-003-98238 Supplier Medchemexpress LLC Supplier No. HYP32445MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-KLVFFA is an inhibitor of Amyloid-β assembly, with an IC50 of 2.6 μM, that can be utilized in Alzheimer's disease studies. This peptide is provided as a white to off-white solid with high purity, making it suitable for various research applications.

  • Inhibitor of Amyloid-β assembly
  • IC50 of 2.6 μM
  • Useful for Alzheimer's disease studies
  • Provided as a white to off-white solid
  • Purity of 99.26%
  • Molecular formula C38H57N7O7
  • Molecular weight of 723.90

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.