Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-KLVFFA | 342877-55-8 | 99.3% | 723.90 | 25 MG
SDP

Catalog No. 5000399006
Encompass_Preferred

D-KLVFFA is an inhibitor of Amyloid-β assembly, with an IC50 of 2.6 μM, that can be used in Alzheimer's disease studies.

  • Inhibitor of Amyloid-β assembly
  • IC50 of 2.6 μM
  • Used in Alzheimer's disease studies
  • Molecular formula: C38H57N7O7
  • Molecular weight: 723.90
  • Sequence: D-Lys-D-Leu-D-Val-D-Phe-D-Phe-D-Ala
  • Appearance: White to off-white solid

Catalog No. 50-003-99006 Supplier Medchemexpress LLC Supplier No. 25MGHYP3244
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-KLVFFA is an inhibitor of Amyloid-β assembly, with an IC50 of 2.6 μM, that can be used in Alzheimer's disease studies.

  • Inhibitor of Amyloid-β assembly
  • IC50 of 2.6 μM
  • Used in Alzheimer's disease studies
  • Molecular formula: C38H57N7O7
  • Molecular weight: 723.90
  • Sequence: D-Lys-D-Leu-D-Val-D-Phe-D-Phe-D-Ala
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.