Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-kynurenine | 13441-51-5 | MFCD00133224 | 99.9% | 208.21 g/mol | C10H12N2O3 | 1 ML
SDP

Catalog No. 5000302127
Encompass_Preferred

D-kynurenine is a research-grade metabolite of D-tryptophan that functions as a GPR109B agonist and a substrate for D-amino acid oxidase. It is supplied as a powder and as a ready-to-use aqueous solution (10 mM, 1 mL) for biochemical and pharmacological assays. The compound has CAS number 13441-51-5 and includes stability guidance for both powder and solvent forms.

  • High purity: 99.94%.
  • Available as 10 mM solution in water, 1 mL.
  • Also available as solid in 25 mg-500 mg pack sizes.
  • Useful as a GPR109B agonist and enzymatic assay substrate.
  • Molecular formula C10H12N2O3; molecular weight 208.21 g/mol.
  • Storage guidance provided for powder and in-solution conditions.

Catalog No. 50-003-02127 Supplier Medchemexpress LLC Supplier No. HYW01450210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-kynurenine is a research-grade metabolite of D-tryptophan that functions as a GPR109B agonist and a substrate for D-amino acid oxidase. It is supplied as a powder and as a ready-to-use aqueous solution (10 mM, 1 mL) for biochemical and pharmacological assays. The compound has CAS number 13441-51-5 and includes stability guidance for both powder and solvent forms.

  • High purity: 99.94%.
  • Available as 10 mM solution in water, 1 mL.
  • Also available as solid in 25 mg-500 mg pack sizes.
  • Useful as a GPR109B agonist and enzymatic assay substrate.
  • Molecular formula C10H12N2O3; molecular weight 208.21 g/mol.
  • Storage guidance provided for powder and in-solution conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.