Learn More
Medchemexpress LLC HY-W014502 25mg Medchemexpress, D-Kynurenine CAS:13441-51-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01450225MG
Encompass_Preferred
Medchemexpress, HY-W014502 25mg D-Kynurenine CAS:13441-51-5 D-kynurenine, a metabolite of D-tryptophan, can serve as the bioprecursor of kynurenic acid (KYNA) and 3-hydroxykynurenine. D-Kynurenine is an agonist for G protein-coupled receptor, GPR109B. D-Kynurenine is a substrate in a fluorometric assay of D-amino acid oxidase. D-kynurenine promotes epithelial-to-mesenchymal transition via activating aryl hydrocarbon receptor (AHR)[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3363