Learn More
Medchemexpress LLC D L-erythro-PDMP hyd 10mg | 80943-40-4 | 427.02 g·mol⁻1 | C23H39ClN2O3 | 10 MG
SDP
Catalog No. 5000308084
Encompass_Preferred
D,L-erythro-PDMP hydrochloride is the erythro stereoisomer of PDMP supplied as a hydrochloride salt for laboratory research. It is used as a biochemical tool to inhibit UDP-glucose:ceramide glucosyltransferase and to study sphingolipid metabolism and cell growth effects in vitro.
  • Inhibits UDP-glucose:ceramide glucosyltransferase (glucosylceramide synthase).
  • Erythro stereoisomer of PDMP.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity (~98.45%) suitable for research use.
  • Provided in a 10 mg research quantity for in vitro studies.
  • Recommended storage: -20°C for the solid; in solvent, -80°C for long-term storage.
Catalog No. 50-003-08084 Supplier Medchemexpress LLC Supplier No. HY116392H10MG
D,L-erythro-PDMP hydrochloride is the erythro stereoisomer of PDMP supplied as a hydrochloride salt for laboratory research. It is used as a biochemical tool to inhibit UDP-glucose:ceramide glucosyltransferase and to study sphingolipid metabolism and cell growth effects in vitro.
  • Inhibits UDP-glucose:ceramide glucosyltransferase (glucosylceramide synthase).
  • Erythro stereoisomer of PDMP.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity (~98.45%) suitable for research use.
  • Provided in a 10 mg research quantity for in vitro studies.
  • Recommended storage: -20°C for the solid; in solvent, -80°C for long-term storage.