Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D L-erythro-PDMP hyd 5mg | 80943-40-4 | 5 MG
SDP

Catalog No. 5000227984
Encompass_Preferred

D,L-erythro-PDMP (hydrochloride) is the erythro stereoisomer mixture of PDMP supplied as the monohydrochloride salt. It is used in biochemical research to modulate glycosphingolipid metabolism and ceramide levels, and has reported effects on glycosylceramide and galactosylceramide pathways; it may inhibit cell growth in vitro at micromolar concentrations.

  • Hydrochloride salt in solid form for laboratory use.
  • Molecular weight ~427.0 g·mol⁻¹.
  • Chemical formula reported as C23H38N2O3 · HCl.
  • Reported purity approximately 98.5%.
  • Typical package size: 5 mg.
  • Modulates glycosphingolipid metabolism and can affect ceramide levels in cells.

Catalog No. 50-002-27984 Supplier Medchemexpress LLC Supplier No. HY116392H5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

D,L-erythro-PDMP (hydrochloride) is the erythro stereoisomer mixture of PDMP supplied as the monohydrochloride salt. It is used in biochemical research to modulate glycosphingolipid metabolism and ceramide levels, and has reported effects on glycosylceramide and galactosylceramide pathways; it may inhibit cell growth in vitro at micromolar concentrations.

  • Hydrochloride salt in solid form for laboratory use.
  • Molecular weight ~427.0 g·mol⁻¹.
  • Chemical formula reported as C23H38N2O3 · HCl.
  • Reported purity approximately 98.5%.
  • Typical package size: 5 mg.
  • Modulates glycosphingolipid metabolism and can affect ceramide levels in cells.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.