Learn More
Cederlane D-/L-Lactic Acid Assay Kit, 100 assays, Enzymatic, Spectrophotometric, Absorbance at 340 nm, 0.5 to 30 μg D- or L-lactic acid range
SDP
Catalog No. NC1382676
Encompass
D-/L-Lactic Acid Assay Kit, 100 assays, Enzymatic, Spectrophotometric, Absorbance at 340 nm, 0.5 to 30 μg D- or L-lactic acid range
Catalog No. NC1382676 Supplier Cederlane Supplier No. 700004276
D-/L-Lactic Acid Assay Kit, 100 assays, Enzymatic, Spectrophotometric, Absorbance at 340 nm, 0.5 to 30 μg D- or L-lactic acid range