Learn More
Medchemexpress LLC d,l-SKF89976A hydrochloride | 85375-15-1 | 98.7% | 371.90 | C22H26ClNO2 | 100 MG
SDP
Catalog No. 5000215846
Encompass_Preferred
SKF89976A hydrochloride is a research reagent that selectively inhibits the GABA transporter GAT-1 and is used in preclinical studies to probe GABA uptake and transporter pharmacology. Reported data in CHO cells show submicromolar potency at GAT-1 and substantially weaker activity at other GAT subtypes.
Catalog No. 50-002-15846 Supplier Medchemexpress LLC Supplier No. HY100228A100MG
SKF89976A hydrochloride is a research reagent that selectively inhibits the GABA transporter GAT-1 and is used in preclinical studies to probe GABA uptake and transporter pharmacology. Reported data in CHO cells show submicromolar potency at GAT-1 and substantially weaker activity at other GAT subtypes.