Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D,L-tryptophanamide hydrochloride | 67607-61-8 | MFCD00070409 | 98.0% | 239.7 g/mol | C11H14ClN3O | 250 MG
SDP

Catalog No. 5000280930
Encompass_Preferred

D,L-Tryptophanamide hydrochloride is the hydrochloride salt of the racemic tryptophanamide derivative, supplied as a research-grade biochemical assay reagent for laboratory use. It is typically provided as a crystalline solid with characterized identity and composition for analytical and assay applications.

  • Used as a biochemical assay reagent for research applications.
  • Racemic tryptophanamide hydrochloride (hydrochloride salt).
  • CAS number 67607-61-8.
  • Molecular weight 239.7 g/mol.
  • Typical purity 98.0%.
  • Available in small laboratory pack sizes (100 mg-1 g).

Catalog No. 50-002-80930 Supplier Medchemexpress LLC Supplier No. HYW343816250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D,L-Tryptophanamide hydrochloride is the hydrochloride salt of the racemic tryptophanamide derivative, supplied as a research-grade biochemical assay reagent for laboratory use. It is typically provided as a crystalline solid with characterized identity and composition for analytical and assay applications.

  • Used as a biochemical assay reagent for research applications.
  • Racemic tryptophanamide hydrochloride (hydrochloride salt).
  • CAS number 67607-61-8.
  • Molecular weight 239.7 g/mol.
  • Typical purity 98.0%.
  • Available in small laboratory pack sizes (100 mg-1 g).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.