Learn More
Medchemexpress LLC D,L-tryptophanamide hydrochloride | 67607-61-8 | MFCD00070409 | 99.1% | 239.70 g/mol | C11H14ClN3O | 100 MG
SDP
Catalog No. 5000282201
Encompass_Preferred
D,L-Tryptophanamide hydrochloride is the hydrochloride salt of racemic tryptophanamide used as a biochemical assay reagent and synthetic building block in research settings. It is provided for research use only and is characterized by standard analytical methods.
  • High purity (99.1% by HPLC).
  • Identity confirmed by LC-MS and spectroscopic analysis.
  • Suitable for use in biochemical assays and small-scale synthesis.
  • Racemic D,L form of tryptophanamide as the hydrochloride salt.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-82201 Supplier Medchemexpress LLC Supplier No. HYW343816100MG
D,L-Tryptophanamide hydrochloride is the hydrochloride salt of racemic tryptophanamide used as a biochemical assay reagent and synthetic building block in research settings. It is provided for research use only and is characterized by standard analytical methods.
  • High purity (99.1% by HPLC).
  • Identity confirmed by LC-MS and spectroscopic analysis.
  • Suitable for use in biochemical assays and small-scale synthesis.
  • Racemic D,L form of tryptophanamide as the hydrochloride salt.
  • Intended for research use only; not for human or clinical use.