Learn More
Cayman Chemical D L1ACtoxychavIcol ACTAt 10mg
SDP
Catalog No. 502984466
Encompass_Preferred
A natural inhibitor of exportin 1, preventing the export of proteins from the nucleus; reduces several intracellular signaling pathways, including NF-κB, impacting inflammation, cancer, viral replication, and other processes
Catalog No. 50-298-4466 Supplier Cayman Chemical Supplier No. 1964910 MG
A natural inhibitor of exportin 1, preventing the export of proteins from the nucleus; reduces several intracellular signaling pathways, including NF-κB, impacting inflammation, cancer, viral replication, and other processes