Learn More
Sigma Aldrich Fine Chemicals Biosciences D Lactate Assay Kit suffic
Catalog No. 502904703
Encompass
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. Monitoring lactate levels is therefore a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. D-lactate is produced in only minor quantities in animals and measuring for D-lactate in animal samples is a means to determine the presence of bacterial infection. Simple direct and automation-ready procedures for measuring lactate concentration are very desirable.
Catalog No. 50-290-4703 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3361KT
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. Monitoring lactate levels is therefore a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. D-lactate is produced in only minor quantities in animals and measuring for D-lactate in animal samples is a means to determine the presence of bacterial infection. Simple direct and automation-ready procedures for measuring lactate concentration are very desirable.