Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-lactate dehydrogenase, microorganism (D-LDH) | 9028-36-8 | ≥800 U/mg protein | 1 KU
SDP

Catalog No. 5000398705
Encompass_Preferred

D-Lactate dehydrogenase, Microorganism (D-LDH) is an oxidoreductase that uses NAD+ or NADP+ as an acceptor and acts on the donor CH-OH group. It catalyzes the oxidation of D-lactate to pyruvate. Widely found in bacteria and fungi, it is often used in biochemical research.

  • Isoelectric point: 4.0
  • Michaelis constant: 1.6x10-4 M (pyruvate, pH 7.0)
  • Inhibitors: Ag+, Hg2+, SH-reagents
  • Optimum pH: 6.0-7.0
  • Optimum temperature: 35-40°C
  • pH stability: 5.0-9.0
  • Appearance: Solid
  • Color: White to off-white
  • EC number: 1.1.1.28
  • Unit definition: One unit will convert one micromole of pyruvate to D-lactate per min at pH 7.4 at 25°C.

Catalog No. 50-003-98705 Supplier Medchemexpress LLC Supplier No. HYP28971KU
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Lactate dehydrogenase, Microorganism (D-LDH) is an oxidoreductase that uses NAD+ or NADP+ as an acceptor and acts on the donor CH-OH group. It catalyzes the oxidation of D-lactate to pyruvate. Widely found in bacteria and fungi, it is often used in biochemical research.

  • Isoelectric point: 4.0
  • Michaelis constant: 1.6x10-4 M (pyruvate, pH 7.0)
  • Inhibitors: Ag+, Hg2+, SH-reagents
  • Optimum pH: 6.0-7.0
  • Optimum temperature: 35-40°C
  • pH stability: 5.0-9.0
  • Appearance: Solid
  • Color: White to off-white
  • EC number: 1.1.1.28
  • Unit definition: One unit will convert one micromole of pyruvate to D-lactate per min at pH 7.4 at 25°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.