Learn More
Medchemexpress LLC D-(-)-Lactic acid-13C | 2102426-04-8 | 99.9% | 91.07 | 50 MG
SDP
Catalog No. 5000343914
Encompass_Preferred
D-(-)-Lactic acid-13C ((R)-2-Hydroxypropionic acid-13C) is a 13C-labeled D-Lactic acid. It can be utilized as an internal standard and for studies involving metabolic tracing.
  • This compound can be used as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-003-43914 Supplier Medchemexpress LLC Supplier No. HY111095S1100MG
D-(-)-Lactic acid-13C ((R)-2-Hydroxypropionic acid-13C) is a 13C-labeled D-Lactic acid. It can be utilized as an internal standard and for studies involving metabolic tracing.
  • This compound can be used as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.