Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(-)-Lactic acid-13C | 2102426-04-8 | 99.9% | 91.07 | C3H6O3 (13C-labeled) | 5 MG
SDP

Catalog No. 5000264518
Encompass_Preferred

D-(-)-Lactic acid-13C is the 13C-labeled D-enantiomer of lactic acid supplied as a solid for laboratory and analytical applications, including tracer and metabolic studies. CAS 2102426-04-8; molecular weight 91.07.

  • Isotopically labeled with 13C for tracer and isotope-ratio studies.
  • High purity (99.9%) suitable for analytical use.
  • Solid, white to off-white form for easy handling and storage.
  • Molecular formula C3H6O3 (13C-labeled) and molecular weight 91.07.
  • Available in small, precise package sizes for research use.

Catalog No. 50-002-64518 Supplier Medchemexpress LLC Supplier No. HY111095S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(-)-Lactic acid-13C is the 13C-labeled D-enantiomer of lactic acid supplied as a solid for laboratory and analytical applications, including tracer and metabolic studies. CAS 2102426-04-8; molecular weight 91.07.

  • Isotopically labeled with 13C for tracer and isotope-ratio studies.
  • High purity (99.9%) suitable for analytical use.
  • Solid, white to off-white form for easy handling and storage.
  • Molecular formula C3H6O3 (13C-labeled) and molecular weight 91.07.
  • Available in small, precise package sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.