Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem D-Lactic acid | 95% | CAS 10326-41-7 | C3H6O3 | 1kg

Catalog No. 5001022629
Encompass_Preferred

D-Lactic acid | 95% | CAS 10326-41-7 | C3H6O3 | 1kg

Catalog No. 50-010-22629 Supplier Aobchem Supplier No. A10199191KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Lactic acid | 95% | CAS 10326-41-7 | C3H6O3 | 1kg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.