Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(-)-Lactic acid (sodium) | 920-49-0 | 98.0% | 112.06 | 10 MG
SDP

Catalog No. 5000343662
Encompass_Preferred

D-(-)-Lactic acid sodium is an endogenous metabolite and is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month.

Catalog No. 50-003-43662 Supplier Medchemexpress LLC Supplier No. HY111095B500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(-)-Lactic acid sodium is an endogenous metabolite and is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.