Learn More
Medchemexpress LLC D-(-)-lactic acid sodium | 920-49-0 | MFCD00066577 | 98.0% | 112.06 g/mol | C3H5NaO3 | 5 MG
SDP
Catalog No. 5000263666
Encompass_Preferred
D-(-)-Lactic acid sodium (sodium D-lactate; CAS 920-49-0) is the sodium salt of the D-enantiomer of lactic acid. It is used as an endogenous metabolite standard and as a reagent in biochemical and metabolic-tracing studies. The compound is supplied as a solid in sealed vials and is reported at 98.0% purity by NMR.
  • Sodium salt form of D-lactic acid, water-soluble and easy to prepare.
  • Suitable as an endogenous metabolite standard for analytical methods.
  • Useful for metabolic tracing and biochemical studies.
  • Reported molecular formula C3H5NaO3 and molecular weight 112.06 g/mol.
  • Purity reported as 98.0% by NMR.
  • Supplied in sealed vial for convenient storage and handling.
Catalog No. 50-002-63666 Supplier Medchemexpress LLC Supplier No. HY111095B5MG
D-(-)-Lactic acid sodium (sodium D-lactate; CAS 920-49-0) is the sodium salt of the D-enantiomer of lactic acid. It is used as an endogenous metabolite standard and as a reagent in biochemical and metabolic-tracing studies. The compound is supplied as a solid in sealed vials and is reported at 98.0% purity by NMR.
  • Sodium salt form of D-lactic acid, water-soluble and easy to prepare.
  • Suitable as an endogenous metabolite standard for analytical methods.
  • Useful for metabolic tracing and biochemical studies.
  • Reported molecular formula C3H5NaO3 and molecular weight 112.06 g/mol.
  • Purity reported as 98.0% by NMR.
  • Supplied in sealed vial for convenient storage and handling.