Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Lactose

Catalog No. AAJ66376A1
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66376A1 1 kg
AAJ6637636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66376A1 Supplier Thermo Scientific Chemicals Supplier No. J66376A1
Add to Cart

D-Lactose, CAS # 63-42-3, is a disaccharide and the major sugar found in milk. It is composed of galactose and glucose subunits. | CAS: 63-42-3 | C12H22O11

D-Lactose is used for the culture of lactic acid metabolizing bacteria, lactic acid bacteria. D-Lactose is used to identify and characterized galectins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D-Lactose is the major sugar found in milk
  • It is formed through the condensation of galactose and glucose

Application

  • D-Lactose is used to identify and characterize galectins
  • It is also used to culture lactic acid-metabolizing bacteria
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-42-3
Molecular Formula C12H22O11
MDL Number MFCD00151251
Synonym Gal-beta-1,4-Glc; 4-O-beta-D-Galactopyranosyl-D-glucopyranose

Specifications

Color White
Odor Odorless
Quantity 1 kg
Beilstein 93796
Merck Index 14,5343
Solubility Information Soluble in water.
Formula Weight 342.3
Physical Form Powder
Chemical Name or Material D-Lactose
EINECSNumber 200-559-2
RTECSNumber OD9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.