Learn More
Alkali Scientific D Lactose Monohydrate, 1 Kg
SDP
Catalog No. 5000660220
Encompass_Preferred
D-Lactose Monohydrate is a high-quality carbohydrate compound used primarily in fermentation applications. This 1 kg reagent is used as a carbon source in microbiological studies and fermentation processes. Lactose Monohydrate is essential for the growth of lactose-fermenting bacteria and fungi. It is commonly utilized in laboratory-scale experiments to analyze enzyme activity and metabolic pathways involving lactose. This reagent is also used in the production of dairy products and in the creation of culture media for microorganism cultivation, particularly for fermenting and testing lactose-metabolizing organisms.
Catalog No. 50-006-60220 Supplier Alkali Scientific Supplier No. DLA001CP
D-Lactose Monohydrate is a high-quality carbohydrate compound used primarily in fermentation applications. This 1 kg reagent is used as a carbon source in microbiological studies and fermentation processes. Lactose Monohydrate is essential for the growth of lactose-fermenting bacteria and fungi. It is commonly utilized in laboratory-scale experiments to analyze enzyme activity and metabolic pathways involving lactose. This reagent is also used in the production of dairy products and in the creation of culture media for microorganism cultivation, particularly for fermenting and testing lactose-metabolizing organisms.