Learn More
Research Products International Corp D-Lactose Monohydrate, 1 KG
SDP
Supplier:  Research Products International Corp L261001000.0
Encompass_Preferred
Lactose monohydrate can be used in bacterial cultures. It is commonly used as a carbon source in bacterial growth media, especially for lactose-fermenting bacteria like E. coli (lac operon system).
Additionally, it is often used in MacConkey agar and Luria-Bertani (LB) broth with lactose for bacterial selection and differentiation.
Catalog No. 50-197-8123