Learn More
Alkali Scientific D Lactose Monohydrate, 5 Kg
SDP
Catalog No. 5000660962
Encompass_Preferred
D-Lactose Monohydrate is a high-purity carbohydrate used in fermentation and microbiological studies. This 5 kg quantity is ideal for large-scale experiments that require a consistent and reliable carbon source for lactose-fermenting microorganisms. The monohydrate form of lactose is often used in media preparation for bacterial and yeast growth, particularly in fermentation processes. It is commonly utilized in the dairy industry for lactose conversion processes, as well as in laboratory research related to microbial metabolism and enzyme activity involving carbohydrates.
Catalog No. 50-006-60962 Supplier Alkali Scientific Supplier No. DLA003CP
D-Lactose Monohydrate is a high-purity carbohydrate used in fermentation and microbiological studies. This 5 kg quantity is ideal for large-scale experiments that require a consistent and reliable carbon source for lactose-fermenting microorganisms. The monohydrate form of lactose is often used in media preparation for bacterial and yeast growth, particularly in fermentation processes. It is commonly utilized in the dairy industry for lactose conversion processes, as well as in laboratory research related to microbial metabolism and enzyme activity involving carbohydrates.