Learn More
Medchemexpress LLC D-Lactose monohydrate | 64044-51-5 | 99.5% | 1 ML
SDP
Catalog No. 5000413669
Encompass_Preferred
D-Lactose monohydrate is a sugar utilized by most bacteria, promoting the growth of tobacco B6S3 tumor tissue. It is also used in research on plant tumor tissue growth mechanisms and bacterial sugar utilization. In vitro, it optimally promotes B6S3 tumor tissue growth at 14.6 mM when cultured for 12 days at concentrations of 2.9-87.5 mM.
  • Utilized by most bacteria
  • Promotes the growth of tobacco B6S3 tumor tissue
  • Used in research on plant tumor tissue growth mechanisms
  • Used in research on bacterial sugar utilization
Catalog No. 50-004-13669 Supplier Medchemexpress LLC Supplier No. HYB167310MM/1ML
D-Lactose monohydrate is a sugar utilized by most bacteria, promoting the growth of tobacco B6S3 tumor tissue. It is also used in research on plant tumor tissue growth mechanisms and bacterial sugar utilization. In vitro, it optimally promotes B6S3 tumor tissue growth at 14.6 mM when cultured for 12 days at concentrations of 2.9-87.5 mM.
  • Utilized by most bacteria
  • Promotes the growth of tobacco B6S3 tumor tissue
  • Used in research on plant tumor tissue growth mechanisms
  • Used in research on bacterial sugar utilization