Learn More
D-(+)-Lactose, Monohydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor ProductsSpecifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Specific Gravity | 1L = 1.53kg |
| Formula Weight | 360.32 |
| Percent Purity | 90 to 100% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Physical Form | Powder |
| Chemical Name or Material | D-(+)-Lactose, Monohydrate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.