Learn More
Medchemexpress LLC D-Lactose monohydrate (Standard) | 64044-51-5 | 99.5% | 500 MG
SDP
Catalog No. 5000411827
Encompass_Preferred
D-Lactose monohydrate is an analytical standard intended for research and analytical applications. This sugar can be utilized by most bacteria and promotes the growth of tobacco B6S3 tumor tissue, making it suitable for research into plant tumor growth mechanisms and bacterial sugar utilization.
  • Analytical and reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: saccharides, polysaccharides
  • Initial source: endogenous metabolite
  • Suitable for research and analytical applications
  • Provides a reference standard for various analytical experiments
Catalog No. 50-004-11827 Supplier Medchemexpress LLC Supplier No. HYB1673500MG
D-Lactose monohydrate is an analytical standard intended for research and analytical applications. This sugar can be utilized by most bacteria and promotes the growth of tobacco B6S3 tumor tissue, making it suitable for research into plant tumor growth mechanisms and bacterial sugar utilization.
  • Analytical and reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: saccharides, polysaccharides
  • Initial source: endogenous metabolite
  • Suitable for research and analytical applications
  • Provides a reference standard for various analytical experiments