Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Lactose Monohydrate

Catalog No. AAJ18185A7 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ18185A7 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ18185A7 Supplier Thermo Scientific Supplier No. J18185A7
Add to Cart

CAS: 64044-51-5 | C12H24O12 | 360.312 g/mol

 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
MDL Number MFCD00150747
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3 S,6 R)-2-(hydroxymethyl)-6-[(3 R,6 R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Melting Point ∼218°C (decomposition)
Density 1.525
Packaging Plastic bottle
Quantity 5 kg
Beilstein 3768231
Specific Gravity 1.525
Formula Weight 360.32
Physical Form Powder
Chemical Name or Material D-(+)-Lactose monohydrate
EINECSNumber 200-559-2
RTECSNumber OD9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.