Learn More
BIOHIPPO D-Lactyllysine Rabbit mAb
SDP
Catalog No. 5000975578
Encompass
U-Blot Pan-D-Lactyllysine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes D-Lactyllysine a lysine lactylation modification at Lys(K) D-Lactyllysine (KD-la) is the D-stereoisomer of lysine lactylation a novel type of lysine acylation post-translational modification By conjugating a D-lactyl group to lysine residues KD-la is mainly generated via non-enzymatic pathways from glycolysis byproducts and exists on both histone and non-histone proteins It participates in chromatin remodeling gene transcription and cellular metabolic stress responses and is closely associated with tumorigenesis inflammation metabolic reprogramming and other pathological processes serving as a key target in metabolic-epigenetic crosstalk research Supplied as 100 uL
Catalog No. 50-009-75578 Supplier BIOHIPPO Supplier No. BHA21821293
U-Blot Pan-D-Lactyllysine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes D-Lactyllysine a lysine lactylation modification at Lys(K) D-Lactyllysine (KD-la) is the D-stereoisomer of lysine lactylation a novel type of lysine acylation post-translational modification By conjugating a D-lactyl group to lysine residues KD-la is mainly generated via non-enzymatic pathways from glycolysis byproducts and exists on both histone and non-histone proteins It participates in chromatin remodeling gene transcription and cellular metabolic stress responses and is closely associated with tumorigenesis inflammation metabolic reprogramming and other pathological processes serving as a key target in metabolic-epigenetic crosstalk research Supplied as 100 uL