Learn More
Medchemexpress LLC D-Leu-Thr-Arg-pNA | 122630-72-2 | 508.57 | 5 MG
SDP
Catalog No. 5000399603
Encompass_Preferred
D-Leu-Thr-Arg-pNA is a substrate for γ-Tryptase, utilized to measure the effects of small molecule inhibitors on γ-Tryptase activity.
  • Substrate for γ-Tryptase
  • Measures effects of small molecule inhibitors on γ-Tryptase activity
  • Target: Ser/Thr Protease
  • Pathway: Metabolic Enzyme/Protease
Catalog No. 50-003-99603 Supplier Medchemexpress LLC Supplier No. HYP44625MG
D-Leu-Thr-Arg-pNA is a substrate for γ-Tryptase, utilized to measure the effects of small molecule inhibitors on γ-Tryptase activity.
  • Substrate for γ-Tryptase
  • Measures effects of small molecule inhibitors on γ-Tryptase activity
  • Target: Ser/Thr Protease
  • Pathway: Metabolic Enzyme/Protease