Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Leu-Thr-Arg-pNA | 122630-72-2 | MFCD02259649 | 508.57 | 10 MG
SDP

Catalog No. 5000397677
Encompass_Preferred

D-Leu-Thr-Arg-pNA is a chemical compound used as a substrate for γ-Tryptase. It can be utilized to measure the effects of small molecule inhibitors on γ-Tryptase activity. This product is intended for testing and research purposes only and is not for use as pharmaceuticals, food, or household goods.

  • Substrate for γ-Tryptase
  • Can be used to measure effects of small molecule inhibitors on γ-Tryptase activity
  • Soluble in DMSO
  • Store at room temperature
  • For testing and research purposes only

Catalog No. 50-003-97677 Supplier Medchemexpress LLC Supplier No. HYP446210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Leu-Thr-Arg-pNA is a chemical compound used as a substrate for γ-Tryptase. It can be utilized to measure the effects of small molecule inhibitors on γ-Tryptase activity. This product is intended for testing and research purposes only and is not for use as pharmaceuticals, food, or household goods.

  • Substrate for γ-Tryptase
  • Can be used to measure effects of small molecule inhibitors on γ-Tryptase activity
  • Soluble in DMSO
  • Store at room temperature
  • For testing and research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.