Learn More
Carbosynth LLC. [D-Leu7]-(-)-Ternatin (1 mg vial), 1ea, MW: 721.97, Molecular formula: C37H67N7O7, CAS# 1005766-32-4
SDP
Supplier:  Carbosynth LLC. TNN4464V1MG
Encompass_Preferred
Synonyms: cyclo(-D-aIle-MeAla-MeLeu-Leu-MeAla-D-MeAla-D-Leu-), Application: Fat Accumulation Inhibitor against 3T3-L1 Adipocytes, Storage Temperature: -20°C, References: K. Shimokawa, I. Mashima, A. Asai, K. Yamada, M. Kita, and D. Uemura, Tetrahedron Lett., 47, 4445 (2006). ((-)-Ternatin; Structure Determination/Biological Activity in vitro), K. Shimokawa, K. Yamada, M. Kita, and D. Uemura, Bioorg. Med. Chem. Lett., 17, 4447 (2007). ((-)-Ternatin; Biological Activity in vivo), M. Ito, J. Ito, H. Kitazawa, K. Shimamura, T. Fukami, S. Tokita, K. Shimokawa, K. Yamada, A. Kanatani and D. Uemura, Peptides, 30, 1074 (2009). ((-)-Ternatin; Mechanism of Inhibitory Activity)
Catalog No. 50-168-7204