Learn More
Medchemexpress LLC HY-Y0378 500mg Medchemexpress, D-Leucine CAS:328-38-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0378500MG
Encompass_Preferred
Medchemexpress, HY-Y0378 500mg D-Leucine CAS:328-38-1 D-Leucine is a more potent anti-seizure agent than L-leucine. D-leucine potently terminates seizures even after the onset of seizure activity. D-leucine, but not L-leucine, reduces long-term potentiation but had no effect on basal synaptic transmission in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3534