Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Leucine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. L01945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L01945G Supplier TCI America Supplier No. L01945G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17664-93-6
Molecular Formula C20H27NO5S
Molecular Weight (g/mol) 393.498
MDL Number MFCD00066116
InChI Key QTQGHKVYLQBJLO-UTONKHPSSA-N
Synonym h-d-leu-obzl.tosoh, d-leucine benzyl ester tosylate, r-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, d-leucine benzyl ester p-toluenesulfonate, h-d-leu-obzl p-tosylate, h-d-leu-obzl tos, d-leucinebenzylestertosylate, benzyl 2r-2-amino-4-methylpentanoate; para-toluene sulfonate, h-d-leu-obzl?os, h-d-leu-obzl.tos
PubChem CID 44629929
IUPAC Name benzyl (2R)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.CC(C)CC(C(=O)OCC1=CC=CC=C1)N

Specifications

Melting Point 158°C
Color White
Quantity 5 g
Formula Weight 393.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Leucine Benzyl Ester p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.