Learn More
Medchemexpress LLC D-leucine-d10 | 271247-12-2 | 99.8% | 141.23 | C6H3D10NO2 | 50mg
SDP
Catalog No. 5000269699
Encompass_Preferred
D-Leucine-d10 is the deuterium labeled D-Leucine D-Leucine is a more potent anti-seizure agent than L-leucine D-leucine potently terminates seizures even after the onset of seizure activity D-leucine but not L-leucine reduces long-term potentiation but had no effect on basal synaptic transmission in vitro[1]
Catalog No. 50-002-69699 Supplier Medchemexpress LLC Supplier No. HYY0378S50MG
D-Leucine-d10 is the deuterium labeled D-Leucine D-Leucine is a more potent anti-seizure agent than L-leucine D-leucine potently terminates seizures even after the onset of seizure activity D-leucine but not L-leucine reduces long-term potentiation but had no effect on basal synaptic transmission in vitro[1]