Learn More
Medchemexpress LLC D-leucine, L-tyrosyl-D-alanylglycyl-L-phenylalanyl- | 63631-40-3 | 99.9% | 569.65 | C29H39N5O7 | 50 MG
SDP
Catalog No. 5000243712
Encompass_Preferred
BW-180C (D-Ala2, D-Leu5)-Enkephalin (DADLE) is a synthetic enkephalin-family peptide that functions as a selective delta-opioid receptor (DOR) agonist for research use. It has been used in cellular and animal studies to probe neuroprotective pathways, modulate transcriptional activity, and assess ischemia-reperfusion injury.
  • Selective delta-opioid receptor agonist for targeted receptor studies.
  • Used in vitro and in vivo to evaluate neuroprotection and ischemia-reperfusion models.
  • Causes reversible inhibition of cellular transcription useful for mechanistic assays.
  • High reported purity (99.9%) supports reproducible experimental results.
  • Molecular weight 569.65 aids dosing and analytical calculations.
  • Available in solid and solution formats for flexible dosing and handling.
Catalog No. 50-002-43712 Supplier Medchemexpress LLC Supplier No. HY10534350MG
BW-180C (D-Ala2, D-Leu5)-Enkephalin (DADLE) is a synthetic enkephalin-family peptide that functions as a selective delta-opioid receptor (DOR) agonist for research use. It has been used in cellular and animal studies to probe neuroprotective pathways, modulate transcriptional activity, and assess ischemia-reperfusion injury.
  • Selective delta-opioid receptor agonist for targeted receptor studies.
  • Used in vitro and in vivo to evaluate neuroprotection and ischemia-reperfusion models.
  • Causes reversible inhibition of cellular transcription useful for mechanistic assays.
  • High reported purity (99.9%) supports reproducible experimental results.
  • Molecular weight 569.65 aids dosing and analytical calculations.
  • Available in solid and solution formats for flexible dosing and handling.