Learn More
Medchemexpress LLC D-Leucine, N-[(1,1-dimethylethoxy) carbonyl]-4-methyl- | 112695-98-4 | 98.9% | C12H23NO4 | 250 MG
SDP
Catalog No. 5000358144
Encompass_Preferred
Boc-beta-t-butyl-d-alanine is an intermediate used in chemical synthesis, particularly for peptides and other amino acids.
  • Used in the synthesis of peptides and other amino acids.
  • Appears as a white to off-white solid.
  • Purity of 98.87%.
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years.
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-58144 Supplier Medchemexpress LLC Supplier No. 250MGHY79404A
Boc-beta-t-butyl-d-alanine is an intermediate used in chemical synthesis, particularly for peptides and other amino acids.
  • Used in the synthesis of peptides and other amino acids.
  • Appears as a white to off-white solid.
  • Purity of 98.87%.
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years.
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month.