Learn More
Medchemexpress LLC D-Leucinol | 53448-09-2 | 99.7% | 117.19 | 10 G
SDP
Catalog No. 5000454852
Encompass_Preferred
D-Leucinol is an isomer of L-Leucinol and can be utilized as an experimental control. L-Leucinol is a competitive aminopeptidase inhibitor with a Ki value of 17 μM. This product is for research use only and not for sale to patients.
  • Isomer of L-Leucinol
  • Can be used as an experimental control
  • Research use only
Catalog No. 50-004-54852 Supplier Medchemexpress LLC Supplier No. HYW01288810G
D-Leucinol is an isomer of L-Leucinol and can be utilized as an experimental control. L-Leucinol is a competitive aminopeptidase inhibitor with a Ki value of 17 μM. This product is for research use only and not for sale to patients.
  • Isomer of L-Leucinol
  • Can be used as an experimental control
  • Research use only