Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Leucinol | 53448-09-2 | 99.7% | 117.19 | 5 G
SDP

Catalog No. 5000454786
Encompass_Preferred

D-Leucinol is an isomer of L-Leucinol and can be used as an experimental control. L-Leucinol is a competitive aminopeptidase inhibitor with a Ki value of 17 μM.

  • Appearance: Liquid (density: 0.917 g/cm³)
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light)

Catalog No. 50-004-54786 Supplier Medchemexpress LLC Supplier No. HYW0128885G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Leucinol is an isomer of L-Leucinol and can be used as an experimental control. L-Leucinol is a competitive aminopeptidase inhibitor with a Ki value of 17 μM.

  • Appearance: Liquid (density: 0.917 g/cm³)
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.