Learn More
Florida Chemical Company d-LimoneneTechnical Grade, 5-Gallon
Supplier:  Florida Chemical Company 3001005GAL
Encompass
This item has a minimum order quantity of 2 per supplier requirements. d-LimoneneTechnical Grade,5-Gallon,35 Pounds Net Weight,Volatile Ingredients: D-Limonene (solvent) is the major component (technical grade 95%) with balance other terpene hydrocarbons and oxygenated compounds- octanal,nonanal,decanal,linalo
Catalog No. NC9213604