Learn More
Medchemexpress LLC HY-112251 100mg Medchemexpress, D-Lin-MC3-DMA CAS:1224606-06-7 Purity:98%
SDP
Catalog No. 501637580
Encompass_Preferred
Medchemexpress,HY-112251 100mg D-Lin-MC3-DMA CAS:1224606-06-7 Formula:C43H79NO2 D-Lin-MC3-DMA, an ionizable cationic lipid, is a potent siRNA delivery vehicle. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7580 Supplier Medchemexpress LLC Supplier No. HY112251100MG
Medchemexpress,HY-112251 100mg D-Lin-MC3-DMA CAS:1224606-06-7 Formula:C43H79NO2 D-Lin-MC3-DMA, an ionizable cationic lipid, is a potent siRNA delivery vehicle. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.