Learn More
Medchemexpress LLC HY-112251 10mg , D-Lin-MC3-DMA CAS:1224606-06-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11225110MG
Encompass_Preferred
HY-112251 10mg D-Lin-MC3-DMA CAS: 1224606-06-7 D-Lin-MC3-DMA, an ionizable cationic lipid, is a potent siRNA delivery vehicle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8962