Learn More
TARGETMOL CHEMICALS INC D-Lin-MC3-DMA 10MG
SDP
Catalog No. 502879998
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Lin-MC3-DMA, an ionizable cationic lipid, serves as a potent siRNA delivery vehicle. Purity 97.18%
Catalog No. 50-287-9998 Supplier TARGETMOL CHEMICALS INC Supplier No. T582310MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Lin-MC3-DMA, an ionizable cationic lipid, serves as a potent siRNA delivery vehicle. Purity 97.18%