Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Lin-MC3-DMA | 1224606-06-7 | 99.9% | 642.09 | 1 ML
SDP

Catalog No. 5000438339
Encompass_Preferred

D-Lin-MC3-DMA is an ionizable cationic lipid that serves as a potent siRNA delivery vehicle. It is intended for research use only and not for sale to patients.

  • Purity: 99.91%
  • Formula: C43H79NO2
  • Appearance: liquid (density: 0.886±0.06 g/cm3), colorless to light yellow
  • Storage: -20°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Functionality: potent siRNA delivery vehicle, especially when incorporated into lipid nanoparticles (LNPs). Its optimized pKa value leads to dramatically enhanced potency in LNP-siRNA systems for gene silencing in hepatocytes.

Catalog No. 50-004-38339 Supplier Medchemexpress LLC Supplier No. HY11225110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Lin-MC3-DMA is an ionizable cationic lipid that serves as a potent siRNA delivery vehicle. It is intended for research use only and not for sale to patients.

  • Purity: 99.91%
  • Formula: C43H79NO2
  • Appearance: liquid (density: 0.886±0.06 g/cm3), colorless to light yellow
  • Storage: -20°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Functionality: potent siRNA delivery vehicle, especially when incorporated into lipid nanoparticles (LNPs). Its optimized pKa value leads to dramatically enhanced potency in LNP-siRNA systems for gene silencing in hepatocytes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.