Learn More
Medchemexpress LLC HY-112251 25mg Medchemexpress, D-Lin-MC3-DMA CAS:1224606-06-7 Purity:>98%
SDP
Catalog No. 502028823
Encompass_Preferred
Medchemexpress, HY-112251 25mg D-Lin-MC3-DMA CAS:1224606-06-7 D-Lin-MC3-DMA, an ionizable amino lipid, is a potent siRNA delivery vehicle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8823 Supplier Medchemexpress LLC Supplier No. HY11225125MG
Medchemexpress, HY-112251 25mg D-Lin-MC3-DMA CAS:1224606-06-7 D-Lin-MC3-DMA, an ionizable amino lipid, is a potent siRNA delivery vehicle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.