Learn More
Selleck Chemical LLC D-Lin-MC3-DMA
SDP
Catalog No. 5000001879
Encompass_Preferred
D-Lin-MC3-DMA (MC3 DLin-MC3-DMA) is a potent and ionizable cationic lipid D-Lin-MC3-DMA is used for delivery of siRNA in vivo
Catalog No. 50-000-01879 Supplier Selleck Chemical LLC Supplier No. S66831G
D-Lin-MC3-DMA (MC3 DLin-MC3-DMA) is a potent and ionizable cationic lipid D-Lin-MC3-DMA is used for delivery of siRNA in vivo