Learn More
Cayman Chemical D-Luciferin, Purity: ≥95%, CAS: 2591-17-5, 100 mg
SDP
Catalog No. 502932848
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-293-2848 Supplier Cayman Chemical Supplier No. 25836100 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP