Learn More
TARGETMOL CHEMICALS INC D-Luciferin 100MG
SDP
Catalog No. 502877825
Encompass_Preferred
Also available in 10 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Luciferin (D-(-)-Luciferin) is a luciferase (Luc) substrate that catalyzes the production of typical yellow-green light (560 nm) by bioluminescent insects. D-Luciferin can be used in luciferase assays. Purity 99.83%
Catalog No. 50-287-7825 Supplier TARGETMOL CHEMICALS INC Supplier No. T3313100MG
Also available in 10 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Luciferin (D-(-)-Luciferin) is a luciferase (Luc) substrate that catalyzes the production of typical yellow-green light (560 nm) by bioluminescent insects. D-Luciferin can be used in luciferase assays. Purity 99.83%