Learn More
Cayman Chemical D-LucIferIn 10mg
SDP
Catalog No. 502933410
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-293-3410 Supplier Cayman Chemical Supplier No. 2583610 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP