Learn More
Selleck Chemical LLC D-Luciferin
SDP
Catalog No. 5000004525
Encompass_Preferred
D-Luciferin is a popular bioluminescent substrate of luciferase in the presence of ATP used in luciferase-based bioluminescence imaging and cell-based high-throughput screening applications In an immunocompetent mouse model of ovarian cancer the use of D-luciferin substrate and firefly luciferase preserves tumour-host immune interactions since bioluminescent imaging is a more sensitive indication of tumour growth than weight gain
Catalog No. 50-000-04525 Supplier Selleck Chemical LLC Supplier No. S776310MM/1ML
D-Luciferin is a popular bioluminescent substrate of luciferase in the presence of ATP used in luciferase-based bioluminescence imaging and cell-based high-throughput screening applications In an immunocompetent mouse model of ovarian cancer the use of D-luciferin substrate and firefly luciferase preserves tumour-host immune interactions since bioluminescent imaging is a more sensitive indication of tumour growth than weight gain