Learn More
Cayman Chemical D-Luciferin, Purity: ≥95%, CAS: 2591-17-5, 250 mg
SDP
Catalog No. 502936781
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-293-6781 Supplier Cayman Chemical Supplier No. 25836250 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP