Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-luciferin | 2591-17-5 | MFCD00042929 | >99.0% | 280.32 g/mol | C11H8N2O3S2 | 5MG
SDP

Catalog No. 5000238042
Encompass_Preferred

D-Luciferin is the natural substrate for luciferase enzymes used in bioluminescent assays and imaging. It produces visible light upon enzymatic oxidation in the presence of ATP, enabling sensitive detection in cell-based and in vivo studies.

  • High purity (≥99.0%) suitable for analytical and biological applications.
  • Effective luciferase substrate for bioluminescent assays and imaging.
  • Soluble in DMSO (125 mg/mL) and sparingly soluble in water unless pH is adjusted.
  • Stable when stored protected from light at 4°C; solutions stable at -80°C for extended storage.
  • Molecular formula C11H8N2O3S2 and molecular weight 280.32 g/mol.

Catalog No. 50-002-38042 Supplier Medchemexpress LLC Supplier No. HY12591A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Luciferin is the natural substrate for luciferase enzymes used in bioluminescent assays and imaging. It produces visible light upon enzymatic oxidation in the presence of ATP, enabling sensitive detection in cell-based and in vivo studies.

  • High purity (≥99.0%) suitable for analytical and biological applications.
  • Effective luciferase substrate for bioluminescent assays and imaging.
  • Soluble in DMSO (125 mg/mL) and sparingly soluble in water unless pH is adjusted.
  • Stable when stored protected from light at 4°C; solutions stable at -80°C for extended storage.
  • Molecular formula C11H8N2O3S2 and molecular weight 280.32 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.